accessibility menu, dialog, popup

UserName

CAS RN 12058-18-3

SeMoSe

CAS RN 12058-18-3

IUPAC Name: diselanylidenemolybdenum
Synonyms: molybdenum diselenide
Molecular Weight (g/mol): 253.89
Molecular Formula: MoSe2
InChi Key: MHWZQNGIEIYAQJ-UHFFFAOYSA-N
SMILES: [Se]=[Mo]=[Se]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results