CAS RN 12057-24-8
CAS RN 12057-24-8
IUPAC Name:
dilithium(1+) hydroxide
Synonyms:
lithium oxide
Molecular Weight (g/mol):
30.89
Molecular Formula:
HLi2O
InChi Key:
XUCJHNOBJLKZNU-UHFFFAOYSA-M
SMILES:
[Li+].[Li+].[OH-]
1
–
1
of
1
results
1
Lithium oxide, 99.5% (metals basis)
CAS: 12057-24-8 Molecular Formula: Li2O Molecular Weight (g/mol): 29.88 MDL Number: MFCD00016183 InChI Key: XZIUWXMPQBQHMC-UHFFFAOYSA-N IUPAC Name: (lithiooxy)lithium SMILES: [Li]O[Li]