accessibility menu, dialog, popup

UserName

CAS RN 12033-81-7

OSOOOPtNNOOOOH3OH3O

CAS RN 12033-81-7

IUPAC Name: sulfuric acid platinum dinitrite
Synonyms: sulfuric acid platinum dinitrite
Molecular Weight (g/mol): 385.17
Molecular Formula: H2N2O8PtS
InChi Key: ZAZPLCLKAKFRMB-UHFFFAOYSA-L
SMILES: [Pt].[O-]N=O.[O-]N=O.OS(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results