CAS RN 1199-46-8
CAS RN 1199-46-8
IUPAC Name:
2-amino-4-tert-butylphenol
Synonyms:
2-amino-4-tert-butylphenol
Molecular Weight (g/mol):
165.24
Molecular Formula:
C10H15NO
InChi Key:
RPJUVNYXHUCRMG-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(O)C(N)=C1
1
–
1
of
1
results
1
2-Amino-4-tert-butylphenol, 97%
CAS: 1199-46-8 Molecular Formula: C10H15NO Molecular Weight (g/mol): 165.24 MDL Number: MFCD00007698 InChI Key: RPJUVNYXHUCRMG-UHFFFAOYSA-N Synonym: 2-amino-4-tert-butyl phenol,2-amino-4-tert-butyl-phenol,2-amino-4-t-butylphenol,unii-28021lmq3d,phenol, 2-amino-4-1,1-dimethylethyl,p-tert-butyl-o-aminophenol,acmc-209a4q,4-tert-butyl-2-aminophenol,bidd:gt0022,2-azanyl-4-tert-butyl-phenol PubChem CID: 70982 IUPAC Name: 2-amino-4-tert-butylphenol SMILES: CC(C)(C)C1=CC=C(O)C(N)=C1