accessibility menu, dialog, popup

UserName

CAS RN 116971-10-9

CH3SBrBr

CAS RN 116971-10-9

IUPAC Name: 2,5-dibromo-3-butylthiophene
Synonyms: 2,5-dibromo-3-butylthiophene
Molecular Weight (g/mol): 298.04
Molecular Formula: C8H10Br2S
InChi Key: YRSQCQUZWSQKKU-UHFFFAOYSA-N
SMILES: CCCCC1=C(Br)SC(Br)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results