accessibility menu, dialog, popup

UserName

CAS RN 116170-84-4

CH3OOSSCH3N

CAS RN 116170-84-4

IUPAC Name: ethyl 4-cyano-5-(methylsulfanyl)thiophene-2-carboxylate
Synonyms: ethyl 4-cyano-5-(methylsulfanyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 227.3
Molecular Formula: C9H9NO2S2
InChi Key: UOHGLSFLWKHJKB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C#N)=C(SC)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results