CAS RN 116070-36-1
CAS RN 116070-36-1
IUPAC Name:
1-(bromomethyl)-3-methyl-5-(trifluoromethyl)benzene
Synonyms:
1-(bromomethyl)-3-methyl-5-(trifluoromethyl)benzene
Molecular Weight (g/mol):
253.06
Molecular Formula:
C9H8BrF3
InChi Key:
GZJBZDJGNUXMNN-UHFFFAOYSA-N
SMILES:
CC1=CC(CBr)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Methyl-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
CAS: 116070-36-1 Molecular Formula: C9H8BrF3 Molecular Weight (g/mol): 253.06 MDL Number: MFCD01631610 InChI Key: GZJBZDJGNUXMNN-UHFFFAOYSA-N Synonym: 1-bromomethyl-3-methyl-5-trifluoromethyl benzene,3-methyl-5-trifluoromethyl benzyl bromide,3-trifluoromethvl-5-methvlbenzvl bromide,3-trifluoromethyl-5-methylbenzyl bromide,benzene, 1-bromomethyl-3-methyl-5-trifluoromethyl PubChem CID: 2775623 IUPAC Name: 1-(bromomethyl)-3-methyl-5-(trifluoromethyl)benzene SMILES: CC1=CC(CBr)=CC(=C1)C(F)(F)F