accessibility menu, dialog, popup

UserName

CAS RN 115029-24-8

IUPAC Name: 2-fluoro-4-(trifluoromethyl)benzoate
Synonyms: 2-fluoro-4-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 207.1
Molecular Formula: C8H3F4O2
InChi Key: OCIYTBZXTFPSPI-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results