accessibility menu, dialog, popup

UserName

H2NO(R)(S)H3N

CAS RN 115014-77-2

IUPAC Name: (1S,2R)-2-carbamoylcyclohexan-1-aminium
Synonyms: (1S,2R)-2-carbamoylcyclohexan-1-aminium
Molecular Weight (g/mol): 143.21
Molecular Formula: C7H15N2O
InChi Key: DXNKJRKPVQVDJW-RITPCOANSA-O
SMILES: NC(=O)[C@@H]1CCCC[C@@H]1[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)