accessibility menu, dialog, popup

UserName

CAS RN 113770-87-9

IUPAC Name: 3-fluoro-4-(trifluoromethyl)pyridine
Synonyms: 3-fluoro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 165.09
Molecular Formula: C6H3F4N
InChi Key: AXHLDFPTUFGBRL-UHFFFAOYSA-N
SMILES: FC1=C(C=CN=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results