accessibility menu, dialog, popup

UserName

CAS RN 1116681-96-9

BOOH3CCH3H3CH3CFClCl

CAS RN 1116681-96-9

IUPAC Name: 2-(4,5-dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4,5-dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 290.95
Molecular Formula: C12H14BCl2FO2
InChi Key: PMLQYOJKCHQKNC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results