CAS RN 109461-46-3
CAS RN 109461-46-3
IUPAC Name:
2-(4-fluorophenyl)-2-methylmorpholine
Synonyms:
2-(4-fluorophenyl)-2-methylmorpholine
Molecular Weight (g/mol):
195.24
Molecular Formula:
C11H14FNO
InChi Key:
IHMLZEWIFPIGKG-UHFFFAOYNA-N
SMILES:
CC1(CNCCO1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
2-(4-Fluorophenyl)-2-methylmorpholine,99%
CAS: 109461-46-3 Molecular Formula: C11H14FNO Molecular Weight (g/mol): 195.237 MDL Number: MFCD08061112 InChI Key: IHMLZEWIFPIGKG-UHFFFAOYSA-N Synonym: 2-4-fluorophenyl-2-methylmorpholine,2-4-fluorophenyl-2-methyl-morpholine,acmc-1c5kc,morpholine,2-4-fluorophenyl-2-methyl PubChem CID: 3066047 IUPAC Name: 2-(4-fluorophenyl)-2-methylmorpholine SMILES: CC1(CNCCO1)C2=CC=C(C=C2)F