CAS RN 1092461-36-3
CAS RN 1092461-36-3
IUPAC Name:
3-bromo-5-(trifluoromethoxy)benzoyl chloride
Synonyms:
3-bromo-5-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol):
303.46
Molecular Formula:
C8H3BrClF3O2
InChi Key:
PRRWHYQCMQADHB-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC(Br)=CC(=C1)C(Cl)=O
1
–
1
of
1
results
1
3-Bromo-5-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™
CAS: 1092461-36-3 Molecular Formula: C8H3BrClF3O2 Molecular Weight (g/mol): 303.46 MDL Number: MFCD11519379 InChI Key: PRRWHYQCMQADHB-UHFFFAOYSA-N Synonym: 3-bromo-5-trifluoromethoxy benzoyl chloride,3-bromo-5-trifluoromethoxy benzoyl chloride, jrd PubChem CID: 54772359 IUPAC Name: 3-bromo-5-(trifluoromethoxy)benzoyl chloride SMILES: FC(F)(F)OC1=CC(Br)=CC(=C1)C(Cl)=O