accessibility menu, dialog, popup

UserName

CAS RN 1092461-18-1

IUPAC Name: 5-chloro-2-(trifluoromethoxy)benzamide
Synonyms: 5-chloro-2-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 239.58
Molecular Formula: C8H5ClF3NO2
InChi Key: NRAWYPLVPCUXOA-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(OC(F)(F)F)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results