CAS RN 1092460-98-4
IUPAC Name:
1-[5-fluoro-2-(trifluoromethoxy)phenyl]methanamine
Synonyms:
1-[5-fluoro-2-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
209.14
Molecular Formula:
C8H7F4NO
InChi Key:
FPZXDOJFLLZXCM-UHFFFAOYSA-N
SMILES:
NCC1=C(OC(F)(F)F)C=CC(F)=C1
1
–
1
of
1
results
1
5-Fluoro-2-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™
CAS: 1092460-98-4 Molecular Formula: C8H7F4NO Molecular Weight (g/mol): 209.14 MDL Number: MFCD11519342 InChI Key: FPZXDOJFLLZXCM-UHFFFAOYSA-N Synonym: 5-fluoro-2-trifluoromethoxy benzylamine,1-5-fluoro-2-trifluoromethoxy phenyl methanamine,5-fluoro-2-trifluoromethoxy benzylamine, jrd,5-fluoro-2-trifluoromethoxy phenyl methylamine PubChem CID: 57365255 IUPAC Name: [5-fluoro-2-(trifluoromethoxy)phenyl]methanamine SMILES: NCC1=C(OC(F)(F)F)C=CC(F)=C1