accessibility menu, dialog, popup

UserName

CAS RN 109010-60-8

CH3CH3H3COONONH2

CAS RN 109010-60-8

IUPAC Name: tert-butyl 2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl)acetate
Synonyms: tert-butyl 2-(3-amino-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl)acetate
Molecular Weight (g/mol): 290.36
Molecular Formula: C16H22N2O3
InChi Key: QTEDVVHLTMELTB-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CN1C2=CC=CC=C2CCC(N)C1=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results