accessibility menu, dialog, popup

UserName

H3COSH2N

CAS RN 105707-24-2

IUPAC Name: 1-(3-amino-5-phenylthiophen-2-yl)ethan-1-one
Synonyms: 1-(3-amino-5-phenylthiophen-2-yl)ethanone
Molecular Weight (g/mol): 217.29
Molecular Formula: C12H11NOS
InChi Key: QAHOJPMILKVSAA-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(N)C=C(S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results