accessibility menu, dialog, popup

UserName

CAS RN 1052694-84-4

NNSH2NFFF

CAS RN 1052694-84-4

IUPAC Name: 5-[2-(trifluoromethyl)phenyl]-1,3,4-thiadiazol-2-amine
Synonyms: 5-[2-(trifluoromethyl)phenyl]-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 245.22
Molecular Formula: C9H6F3N3S
InChi Key: TWXQRAMCZAUPAN-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results