accessibility menu, dialog, popup

UserName

CAS RN 105242-10-2

HNNH

CAS RN 105242-10-2

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}piperazine
Synonyms: 2-{[1,1'-biphenyl]-4-yl}piperazine
Molecular Weight (g/mol): 238.33
Molecular Formula: C16H18N2
InChi Key: QUINYTMXFWCKTN-UHFFFAOYNA-N
SMILES: C1CNC(CN1)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results