accessibility menu, dialog, popup

UserName

CAS RN 10397-13-4

ONNNClCl

CAS RN 10397-13-4

IUPAC Name: 4-(4,6-dichloropyrimidin-2-yl)morpholine
Synonyms: 4-(4,6-dichloropyrimidin-2-yl)morpholine
Molecular Weight (g/mol): 234.08
Molecular Formula: C8H9Cl2N3O
InChi Key: OXCOCPRVQUEIOL-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=NC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results