accessibility menu, dialog, popup

UserName

CAS RN 103877-80-1

CH3FOHOF

CAS RN 103877-80-1

IUPAC Name: 2,5-difluoro-4-methylbenzoic acid
Synonyms: 2,5-difluoro-4-methylbenzoic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: PWWMQUUDAAWEBK-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(C(O)=O)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results