CAS RN 103733-40-0
CAS RN 103733-40-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-tert-Butyl 2-[(S)-2-[(S)-1-ethoxy-1-oxo-4-phenylbutan-2-ylamino]propanoyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylate Maleate (Moexipril tert-Butyl Ester Maleate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.