accessibility menu, dialog, popup

UserName

CAS RN 10366-35-5

H2NONCl

CAS RN 10366-35-5

IUPAC Name: 2-chloropyridine-3-carboxamide
Synonyms: 2-chloropyridine-3-carboxamide
Molecular Weight (g/mol): 156.57
Molecular Formula: C6H5ClN2O
InChi Key: ZQZAHPFFZWEUCL-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CN=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)