accessibility menu, dialog, popup

UserName

ClI

CAS RN 10297-05-9

IUPAC Name: 1-chloro-4-iodobutane
Synonyms: 1-chloro-4-iodobutane
Molecular Weight (g/mol): 218.46
Molecular Formula: C4H8ClI
InChi Key: JXOSPTBRSOYXGC-UHFFFAOYSA-N
SMILES: ClCCCCI
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results