accessibility menu, dialog, popup

UserName

CAS RN 10129-78-9

IUPAC Name: 2-(2,4-dibromophenoxy)acetate
Synonyms: 2-(2,4-dibromophenoxy)acetate
Molecular Weight (g/mol): 308.93
Molecular Formula: C8H5Br2O3
InChi Key: LIIAWXHMVYLFGT-UHFFFAOYSA-M
SMILES: [O-]C(=O)COC1=CC=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results