Fisherbrand™ Q705 Sonicator with Probe
Use this high powered sonicator for any application and sample volume. Fisherbrand™ Sonicators are ideal for cell disruption, nanoparticle dispersion, DNA shearing/ChIP and homogenization.
Product Details
53600.97 NOK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Additional Details:
Additional Details: Weight: 11.43000kg
Product Code. 12931151
Quantity Price
1 58595.0  NOK / Each
Estimated Shipment
05-05-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Accessories and Spare Parts
Description and Specification
Specification
Voltage 230 V
Electrical Requirements 230V, 50/60Hz
Hertz 20 kHz
Wattage 700 W
Dimensions (L x W x H) 387 x 203 x 216 mm (15.25 x 8 x 8.5 in.)
Model Q705
  • 700W
  • Touch screen control
  • 10 programmable memories
  • Programmable from 1 second to 99 hours
  • Overload protection
  • Temperature monitoring
  • Amplitude control from 1 to 100%
  • Processing volume 0.2mL to 1L
Alternative Products
Documentation Accessories and Spare Parts