Learn More
ZNF469 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567072
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PCPASFHPGHA ALLPCAQEDL VSGAPFSPRG ANFHFQPVQK AGASKTGLCQ AEGDSRPPQD VCLPEPSKQP GPQLDAGSLA KCSPDQELSF Highest antigen sequence identity to the following orthologs - mouse 32%, rat 30%.
May be involved in transcriptional regulation.Specifications
ZNF469 | |
Polyclonal | |
Unconjugated | |
ZNF469 | |
KIAA1858 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84627 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JG9 | |
ZNF469 | |
Recombit Human ZNF469 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only