missing translation for 'onlineSavingsMsg'
Learn More

ZNF276 Antibody (PCRP-ZNF276-1A5) [Janelia Fluor™ 585], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-14066JF585

Product Code. 30171304

  • 6275.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF276 Monoclonal antibody specifically detects ZNF276 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF276
Monoclonal
Janelia Fluor 585
DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF
Recombinant full-length human ZNF276 protein (Uniprot: Q8N554)
0.1 mL
Primary
Human
Purified
Flow Cytometry, ELISA, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence
PCRP-ZNF276-1A5
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
92822
Store at 4°C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.