missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 639 Antibody (PCRP-ZNF639-2B2) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14131JF669
This item is not returnable.
View return policy
Description
Zinc finger protein 639 Monoclonal antibody specifically detects Zinc finger protein 639 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| Zinc finger protein 639 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| ANC_2H01, ZASC1ANC-2H01, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein 639, Zinc finger protein ANC_2H01, Zinc finger protein ZASC1,6230400O18Rik | |
| Recombinant human fragment (aa 406-485) of Zinc finger protein 639 (Uniprot: Q9UID6) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-ZNF639-2B2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 51193 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction