missing translation for 'onlineSavingsMsg'
Learn More

ZBTB1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26287-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894795

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZBTB1 acts as a transcriptional repressor. Represses cAMP-responsive element (CRE)-mediated transcriptional activation. In addition, has a role in translesion DNA synthesis. Requires for UV-inducible RAD18 loading, PCNA monoubiquitination, POLH recruitment to replication factories and efficient translesion DNA synthesis. Plays a key role in the transcriptional regulation of T lymphocyte development.
Specifications

Specifications

ZBTB1
Polyclonal
Unconjugated
ZBTB1
KIAA0997, ZBTB1
Rabbit
Antigen Affinity Chromatography
RUO
22890
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y2K1
ZBTB1
ZBTB1 Fusion Protein Ag23492
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers