Learn More
Invitrogen™ Human YES1, GST Tag Recombinant Protein
YES1 is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
Brand:  Invitrogen™ A15557
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15313816
  • 4705.00 NOK / 10µg
Estimated Shipment: 07-05-2024
to see stock.
Description
Description
  • Active—this protein can be used in kinase activity assays, such as Z’-LYTE™ assays, or in a binding assay platform like the LanthaScreen™ Eu Kinase Binding Assay (KBA).
  • Purified—purity is determined by gel electrophoresis and provided in lot-specific certificate of analysis (COA).
  • GST-tagged—provides animal origin flexibility. When used in the KBA platform, it can be paired with a goat- or rabbit-derived Eu-anti-GST antibody.
Specifications
Specifications
NP_005424
88.3 kDa
YES1
Human
GST Tag
Tyrosine Kinases
Kinases & Inhibitors
Liquid
Kinase Assay
10μg
SRC Kinase Family
Recombinant
50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% glycerol.
For maximum recovery please spin prior to use. Aliquoting the 5 μg, 10 μg, and 20 μg sizes of kinase is not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Never store a kinase diluted. If properly stored at -80°C, this product is guaranteed for 6 months from date of purchase.
Baculovirus
Recombinant, Full Length
Signal Transduction