Learn More
Uvex Xenova™ Micro Black/Red PU/PU 95049 Sandal - Width 12
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1105.00 NOK
Specifications
Color | Black, Red |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17287531
|
Uvex
9504938 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17297531
|
Uvex
9504939 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17207541
|
Uvex
9504940 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17217541
|
Uvex
9504941 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17227541
|
Uvex
9504942 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17237541
|
Uvex
9504943 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17247541
|
Uvex
9504944 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17257541
|
Uvex
9504945 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17267541
|
Uvex
9504946 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17277541
|
Uvex
9504947 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17287541
|
Uvex
9504948 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17297541
|
Uvex
9504949 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17207551
|
Uvex
9504950 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17217551
|
Uvex
9504951 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17227551
|
Uvex
9504952 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Uvex 2 Xenova™ Sandal 9504.9 S1 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side, no thermal conductivity and anti-magnetic properties
- Breathable, high-tech microvelour upper material with large perforations
- Distance mesh lining provides optimum breathability
- Made from synthetic material, suitable for people allergic to chrome
- Double hook and loop fastening system for individual adjustment
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft, padded collar and tongue
- Comfortable removable insole, antistatic with a moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant, free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistant up to 120°C
- Antistatic
- Fulfils ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 9504.9
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 38 to 52
Specifications
Black, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |