Learn More
Uvex Xenova™ Micro Black/Green PU/PU 95597 Sandal - Width 10
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1195.00 NOK
Specifications
Color | Black, Green |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17288631
|
Uvex
9559738 |
38 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17298631
|
Uvex
9559739 |
39 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17208641
|
Uvex
9559740 |
40 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17218641
|
Uvex
9559741 |
41 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17228641
|
Uvex
9559742 |
42 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17238641
|
Uvex
9559743 |
43 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17248641
|
Uvex
9559744 |
44 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17258641
|
Uvex
9559745 |
45 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17268641
|
Uvex
9559746 |
46 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17278641
|
Uvex
9559747 |
47 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17288641
|
Uvex
9559748 |
48 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17298641
|
Uvex
9559749 |
49 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17208651
|
Uvex
9559750 |
50 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17218651
|
Uvex
9559751 |
51 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
17228651
|
Uvex
9559752 |
52 |
1195.00 NOK
1 pair |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Xenova™ Sandal 95597 S1 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toecap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Breathable, high-tech microvelour upper material with large perforations
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for people allergic to chrome
- Double hook and loop fastening system for individual adjustment
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft padded collar and tongue
- Comfortable removable insole, antistatic with moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant
- Free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistance up to 120°C
- Antistatic
- Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 95597
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 38 to 52
Specifications
Black, Green | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S1 |