Learn More
Uvex Xenova™ Black/Red PU 95079 Boot - Width 12
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1445.00 NOK
Specifications
Color | Black, Red |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17237761
|
Uvex
9507938 |
38 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17247761
|
Uvex
9507939 |
39 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17257761
|
Uvex
9507940 |
40 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17267761
|
Uvex
9507941 |
41 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17277761
|
Uvex
9507942 |
42 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17287761
|
Uvex
9507943 |
43 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17297761
|
Uvex
9507944 |
44 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17207771
|
Uvex
9507945 |
45 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17217771
|
Uvex
9507946 |
46 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17227771
|
Uvex
9507947 |
47 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17237771
|
Uvex
9507948 |
48 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17247771
|
Uvex
9507949 |
49 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17257771
|
Uvex
9507950 |
50 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17267771
|
Uvex
9507951 |
51 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17277771
|
Uvex
9507952 |
52 |
1445.00 NOK
1 pair |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Xenova™ Boot 9507.9 S2 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Breathable, water repellent full grain leather
- Distance mesh lining provides optimum breathability
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft, padded collar and dust tongue
- Comfortable removable insole, antistatic with a moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant, free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistant up to 120°C
- Antistatic
- Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 9507.9
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S2 |