Learn More
Uvex Xenova™ Black/Red PU 95077 Boot - Width 10
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1445.00 NOK
Specifications
Color | Black, Red |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17237731
|
Uvex
9507738 |
38 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17247731
|
Uvex
9507739 |
39 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17257731
|
Uvex
9507740 |
40 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17267731
|
Uvex
9507741 |
41 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17277731
|
Uvex
9507742 |
42 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17287731
|
Uvex
9507743 |
43 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17297731
|
Uvex
9507744 |
44 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17207741
|
Uvex
9507745 |
45 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17217741
|
Uvex
9507746 |
46 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17227741
|
Uvex
9507747 |
47 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17247741
|
Uvex
9507749 |
49 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17257741
|
Uvex
9507750 |
50 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17267741
|
Uvex
9507751 |
51 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17277741
|
Uvex
9507752 |
52 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17237741
|
Uvex
9507748 |
48 |
1445.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Xenova™ Boot 9507.7 S2 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Breathable, water repellent full grain leather
- Distance mesh lining provides optimum breathability
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft, padded collar and dust tongue
- Comfortable removable insole, antistatic with a moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant, free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistant up to 120°C
- Antistatic
- Fulfils ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 9507.7
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |