Learn More
Uvex Xenova™ Black/Red PU 95061 Low Shoe - Width 10
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1430.00 NOK
Specifications
Color | Black, Red |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17287591
|
Uvex
9506138 |
38 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17297591
|
Uvex
9506139 |
39 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17207601
|
Uvex
9506140 |
40 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17217601
|
Uvex
9506141 |
41 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17227601
|
Uvex
9506142 |
42 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17237601
|
Uvex
9506143 |
43 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17247601
|
Uvex
9506144 |
44 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17257601
|
Uvex
9506145 |
45 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17267601
|
Uvex
9506146 |
46 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17277601
|
Uvex
9506147 |
47 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17287601
|
Uvex
9506148 |
48 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17297601
|
Uvex
9506149 |
49 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17207611
|
Uvex
9506150 |
50 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17217611
|
Uvex
9506151 |
51 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
17227611
|
Uvex
9506152 |
52 |
1430.00 NOK
1 pair |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Xenova™ Shoe 9506.1 S3 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side, no thermal conductivity and anti-magnetic properties
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
- Breathable, water repellent full grain leather
- Scratch resistant scuffcap made from PU coated leather for longer service life
- Distance mesh lining provides optimum breathability
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft, padded collar and dust tongue
- Comfortable removable insole, antistatic with a moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant, free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistant up to 120°C
- Antistatic
- Fulfils ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 9506.1
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 38 to 52
Specifications
Black, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S3 |