Learn More
DWK Life Sciences Wheaton™ Flat Bottom Flasks For Soxhlet Extraction Apparatus
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
491.00 NOK - 575.00 NOK
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | For continuous extraction of analytes from a solid into an organic solvent |
Standard Taper Joint | 24/40 |
Bottom Shape | Flat |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12447111
|
DWK Life Sciences
428126 |
125 mL |
530.00 NOK
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
12457111
|
DWK Life Sciences
428146 |
250 mL |
491.00 NOK
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
12467111
|
DWK Life Sciences
428164 |
500 mL |
575.00 NOK
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
Description
- Flat bottom flask
Specifications
Borosilicate Glass | |
For continuous extraction of analytes from a solid into an organic solvent | |
24/40 | |
Flat |