missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Purple
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100 Cs.
1000 Cs.
Description
- Packaged in case of 100 or 1000
Specifications
For Use With (Equipment) | Plastic dropping bottle |
Product Type | Polypropylene Cap |
Color | Purple |
Material | Polypropylene |
No. per Case | 100 Cs. |
Neck Finish | 20-410 |
Certifications/Compliance | ISO 9001 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'