Learn More
DWK Life Sciences Wheaton™ Natural Polypropylene Caps for Narrow-Mouth Containers
For use with Wheaton leak-resistant narrow mouth bottles
Brand:  DWK Life Sciences 239506
Additional Details : Weight : 0.47630kg
 View more versions of this product
Product Code. 12473851
  • 336.30 NOK / Pack of 72
Estimated Shipment: 24-02-2022
Description
Description
  • Leak-proof design
  • Autoclavable
Specifications
Specifications
ISO 9001
Natural
72 Cs.
Redi-pak™ leak resistant narrow mouth bottle
Polypropylene
20-410