missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Natural Polypropylene Caps for Narrow-Mouth Containers

Catalog Number p-75583
Click to view available options
Neck Finish:
13-425
20-410
24-410
38-430
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with Wheaton leak-resistant narrow mouth bottles

  • Leak-proof design
  • Autoclavable

For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Color Natural
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'