Learn More
DWK Life Sciences Wheaton™ Natural HDPE Cylinder Rounds with Foamed PE-Lined Caps
Brand:  DWK Life Sciences 221154
Additional Details : Weight : 1.27010kg
 View more versions of this product
Product Code. 12442581
  • 777.69 NOK / Pack of 72
Estimated Shipment: 23-02-2022
Description
Description
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner
Specifications
Specifications
ISO 9001
Polyethylene
Screw Cap
3.85 in.
2 oz., 60mL
2 oz.
1.37 in.
Cylinder Round Bottle
20-410
Polyethylene Foam
20-410 mm
35 mm
98 mm
60 mL
Natural
HDPE