Learn More
DWK Life Sciences Wheaton™ Media/Laboratory Bottle Caps
For use with Wheaton media bottles
Brand:  DWK Life Sciences 240080
Additional Details : Weight : 2.26800kg
 View more versions of this product
Product Code. 12473871
  • 2197.12 NOK / Pack of 200
Estimated Shipment: 24-02-2022
Description
Description
  • Black phenolic caps with screw thread
  • Low Density Polyethylene (Not autoclavable), Rubber lined (Autoclavable)
Specifications
Specifications
Media bottle
Polyethylene
33-430
Black
200 Cs.