Learn More
DWK Life Sciences Wheaton™ LDPE Spray Bottles
Catalog Number 12473701
Click to view available options
Capacity (Metric):
20 mL
35 mL
60 mL
Wheaton Spray Bottle made of white LDPE
  • Bottle made of white LDPE (Low Density Polyethylene)
  • Includes bottle, atomizer tip, dip tube and screw cap
  • Screw cap made from polypropylene
Capacity 1.08 oz., 35mL
Closure Material Polypropylene
Closure Size 18-410 mm
Closure Type Screw Cap
Height (English) 3.7 in.
Height (Metric) 94 mm
Length (English) 0.86 in.
Width (English) 1.81 in.
Product Type Spray Bottle
Capacity (Metric) 35 mL
Capacity (English) 1.08 oz.
Color White
Dimensions (L x W x H) 0.86 x 1.81 x 3.7 in. (22 x 46 x 94mm)
Includes Bottle, atomizer tip, dip tube and screw cap
Material LDPE
Length (Metric) 22mm
Width (Metric) 46mm