Learn More
DWK Life Sciences Wheaton™ Connection Connectors
Safely joins two exterior threaded glass components without hooks, springs or clamps
Brand:  DWK Life Sciences 125488
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 12445442
  • 2720.00 NOK / Pack of 6
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Screw cap configuration creates a grease-free, vacuum- tight seal
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Available with white polypropylene or black phenolic caps
  • Phenolic connectors have a working temperature of up to 180
Specifications
Specifications
Screw Thread Connector
Borosilicate Glass
Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Screw cap
USP Type I, ASTM E438 Type I, Class A
Micro soxhlet extraction apparatus
20-400 and 24-400
Polypropylene