Learn More
DWK Life Sciences™ Wheaton™ Caps for 12 x 32 mm Large Opening ABC Vials
These closures for ABC Vials feature the Wheaton™ Super Septa, which is low extractable and optimal for GC/MS.
Brand: DWK Life Sciences™ W225332-0207
Additional Details : Weight : 0.92990kg
Description
- Screw caps are available in natural, black, red, blue, yellow or green polypropylene
Specifications
Green | |
No | |
100 Caps/Pk., 10 Pk./Cs. |
Polypropylene | |
PTFE, Silicone |