Learn More
DWK Life Sciences Wheaton™ ABC Clear Glass Vial Convenience Packs
40% larger opening improves sample accessibility while reducing autosampler needle damage
725.00 NOK - 1280.00 NOK
Specifications
Diameter (English) Outer | 0.47 in. |
---|---|
Color | Clear |
Diameter (Metric) Outer | 12 mm |
Material | Borosilicate Glass |
Volume (Metric) | 1.8 mL |
Product Code | Brand | Closure Color | Septum | Pre-slit | Marking Spot | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Septum | Pre-slit | Marking Spot | Price | Quantity & Availability | ||
12444891
|
DWK Life Sciences
W225154-0401 |
Black | Blue PTFE, White Silicone | No | No |
1280.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12494821
|
DWK Life Sciences
W225150-0801 |
Black | Red PTFE, White Silicone | Yes | No |
1280.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12484821
|
DWK Life Sciences
W225150-08 |
Natural | Red PTFE, White Silicone | Yes | No |
1280.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12474811
|
DWK Life Sciences
W225150-0201 |
Black | Red PTFE, White Silicone | No | No |
935.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12424821
|
DWK Life Sciences
W225150-03 |
Natural | Red PTFE, White Silicone, Red PTFE | No | No |
1090.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12494811
|
DWK Life Sciences
W225150-0204 |
Blue | Red PTFE, White Silicone | No | No |
935.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12464811
|
DWK Life Sciences
W225150-02 |
Natural | Red PTFE, White Silicone | No | No |
935.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12434891
|
DWK Life Sciences
W225154-04 |
Natural | Blue PTFE, White Silicone | No | No |
905.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12474821
|
DWK Life Sciences
W225150-0601 |
Black | Red PTFE | No | No |
725.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12414821
|
DWK Life Sciences
W225150-0207 |
Green | Red PTFE, White Silicone | No | No |
935.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12434821
|
DWK Life Sciences
W225150-0301 |
Black | Red PTFE, White Silicone, Red PTFE | No | No |
1090.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12464821
|
DWK Life Sciences
W225150-06 |
Natural | Red PTFE | No | No |
730.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
12404821
|
DWK Life Sciences
W225150-0205 |
Yellow | Red PTFE, White Silicone | No | No |
935.00 NOK
Pack of 100 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
Description
- Manufactured from clear Type I borosilicate glass
- 12 x 32 mm screw top with 9 mm ABC screw cap
- Available with and without writing patch
Specifications
0.47 in. | |
12 mm | |
1.8 mL | |
Polypropylene | |
32 mm |
Clear | |
Borosilicate Glass | |
1.25 in. | |
Screw Cap | |
9 mm |