Learn More
Cytiva Whatman™ Pop-Top Plastic Filter Holder
Filter holders designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
Brand: Cytiva 420100
1102.00 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.15000kg
Description
- Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
- All three holders will accommodate Nuclepore track-etched and cast membranes.
- Syringe compatible.
Specifications
Pop-Top Plastic Filter Holder | |
Polypropylene | |
3.7 cm | |
13 mm |