missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 520 a Filter Papers for Technical Use
A thin paper with great wet strength and a very high flow rate. Frequently used in technical applications such as the filtration of viscous liquids and emulsions (e.g., sweetened juices, spirits and syrups, resin solutions, oils, or plant extracts).
Brand: Cytiva 10331487
Additional Details : Weight : 7.60000kg
Description
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.Specifications
Grade 520 a Filter Paper for Technical Use, sheet, 580 × 580 mm | |
18 μm | |
A thin paper with great wet strength and a very high flow rate. Frequently used in technical applications such as the filtration of viscous liquids and emulsions (e.g., sweetened juices, spirits and syrups, resin solutions, oils, or plant extracts). | |
250 Pieces |
Cellulose | |
Approx. 17.5 s | |
Grade 520 a | |
580 x 580 mm (23 x 23 in.) |