Learn More
Eppendorf™ Weighing Vessel
273.00 NOK - 326.00 NOK
Specifications
Type | Weighing Vessel |
---|---|
For Use With (Application) | For Use With Calibration Kit |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10657714
|
Eppendorf™
3113808003 |
15 mm |
273.00 NOK
Pack of 3 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10025833
|
Eppendorf™
3113807007 |
24 mm |
326.00 NOK
Pack of 3 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Specifications
Weighing Vessel | |
For Use With Calibration Kit |