missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22614-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes the full length WDR33 protein.
WDR33 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination.This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
WDR33 | |
Polyclonal | |
Unconjugated | |
WDR33 | |
NET14, WD repeat domain 33, WDC146, WDR33 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55339 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9C0J8 | |
WDR33 | |
WDR33 Fusion Protein Ag18382 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |